Learn More
BRAND™ 24-well Insert 2in1, PC-Membrane
Product Code. 16184947 Shop All Brand Products
Change view
Click to view available options
Pore Size:
0.4 μm
8 μm
Quantity:
4 x 12
48 x 1
Unit Size:
Pack of 48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity Unit Size
16184947 8 μm 48 x 1 Pack of 48
16164947 0.4 μm 48 x 1 Pack of 48
16174947 0.4 μm 4 x 12 Pack of 48
16194947 8 μm 4 x 12 Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16184947 Supplier BRAND™ Supplier No. 782706
  • ONLINE EXCLUSIVE PRICE
    384.00€
    345.85€ / Pack of 48
    Save 38.15€
    9% Off
Estimated Shipment: 09-12-2026
The cellGrade™ plus surface of the membranes ensures excellent cell attachment and growth. The highly transparent membranes give you good visibilty of your cell cultures, and the high pore density contributes to good permeability.
  • Use Inserts in a hanging or standing position
  • For 24-well plates or culture dishes
  • Achieve uniform and controlled cell growth due to surface treatment, without coatings
TRUSTED_SUSTAINABILITY
Specifications
Description 24-well Insert 2in1, PC-Membrane 8 μm
Material Polycarbonate
Pore Size 8 μm
Quantity 48 x 1
Sterility Sterile
No. of Wells 24