Learn More
uvex 2124 Xact-Fit Multi Reusable Ear Plugs with Storage Case
Shop All Uvex Products
Click to view available options
Size:
Regular
Small
Unit Size:
Pack of 50
Description
- Unique oval design adjusts perfectly to the anatomy of the ear canal
- Angled pin significantly reduces pressure in the ear
- Pre-formed pins make it easy to insert and remove ear plugs
- Thumb indentations for enable quick and easy handling
- Smooth, dirt-repellent surface can be cleaned with mild soap and water
- Comfortably wear corded ear plugs around the neck in low-noise areas
- Clear, hygienic box included for safe storage when not in use
Specifications
Specifications
| Color | Transparent, Gray |
| ANSI S3.1974 | No |
| Cord Color | Lime Green |
| Size | Regular |
| For Use With (Application) | Variable Environments |
| Model | 2124 Xact-Fit Multi |
| Noise Reduction Rating (NRR) | 26 dB |
| Corded | Corded |
| Plug Shape | Oval |
| Product Type | Ear Plugs |
| Show More |