Learn More
(20S)-Protopanaxadiol, TRC
Specifications
Specifications
| Chemical Name or Material | (20S)-Protopanaxadiol |
| CAS | 30636-90-9 |
| Molecular Formula | C30H52O3 |
| Quantity | 50 mg |
| Synonym | Dammar-24-ene-3β,12β,20-triol, (20S)-Protopanaxadiol, 20(S)-APPD, 20-Epiprotopanaxadiol, (3β,12β)-Dammar-24-ene-3,12,20-triol |
| InChI Formula | InChI=1S/C30H52O3/c1-19(2)10-9-14-30(8,33)20-11-16-29(7)25(20)21(31)18-23-27(5)15-13-24(32)26(3,4)22(27)12-17-28(23,29)6/h10,20-25,31-33H,9,11-18H2,1-8H3/t20-,21+,22-,23+,24-,25-,27-,28+,29+,30-/m0/s1 |
| SMILES | CC(=CCC[C@@](C)([C@H]1CC[C@]2(C)[C@@H]1[C@@H](C[C@@H]3[C@@]4(C)CC[C@@H](C(C)(C)[C@@H]4CC[C@]32C)O)O)O)C |
| Molecular Weight (g/mol) | 460.73 |
| Formula Weight | 460.3916 |