Learn More
(20S)-21-[[Tris(isopropyll)silyl]oxy]-20-methyl-pregn-4-en-3-one, TRC
Specifications
Specifications
| Chemical Name or Material | (20S)-21-((Tris(isopropyll)silyl)oxy)-20-methyl-pregn-4-en-3-one |
| CAS | 356063-47-3 |
| Molecular Formula | C31H54O2Si |
| Quantity | 100 mg |
| Synonym | (20S)-20-Methyl-21-[[tris(1-methylethyl)silyl]oxy]-pregn-4-en-3-one |
| InChI Formula | InChI=1S/C31H54O2Si/c1-20(2)34(21(3)4,22(5)6)33-19-23(7)27-12-13-28-26-11-10-24-18-25(32)14-16-30(24,8)29(26)15-17-31(27,28)9/h18,20-23,26-29H,10-17,19H2,1-9H3/t23-,26+,27-,28+,29+,30+,31-/m1/s1 |
| SMILES | CC(C)[Si](OC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)(C(C)C)C(C)C |
| IUPAC Name | (8S,9S,10R,13S,14S,17R)-10,13-dimethyl-17-[(2S)-1-tri(propan-2-yl)silyloxypropan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 486.84 |
| Formula Weight | 486.3893 |