Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve Set, ASTM Certificate
Shop All Retsch ProductsSpecifications
Specifications
| Quantity | 1Each |
| Pore Size | 325 mesh, 230 mesh, 120 mesh, 60 mesh, 35 mesh, 18 mesh, 10 mesh, 5 mesh |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E 11 |
| Material | Stainless Steel |
| For Use With (Equipment) | Shaker AS 200 Tap |
| Includes | Certificate indicating Compliance, 8 test sieves (325 mesh, 230 mesh, 120 mesh, 60 mesh, 35 mesh, 18 mesh, 10 mesh, 5 mesh) and collecting pan |
| Height (Metric) | 50.8mm |
| For Use With (Application) | For separation, fractioning, particle size determination |
| Diameter (Metric) Outer | 203.2 mm |
| Show More |