Learn More
Fisher Chemical 200LT DICHLOROMETHANE - CERTIFIED REAGEN
Brand:  Fisher Chemical D/1852/27
200LT DICHLOROMETHANE - CERTIFIED REAGEN
Product Code. 10722454
  • ONLINE EXCLUSIVE PRICE
    1195.00€
    1147.00€ / 200 L
    Save 48.00€
    4% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.