Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Product Code. 15816752 Shop All Ohaus Products
Click to view available options
Capacity:
1 x 100 mL
1 x 250 mL
1 x 50 mL (29 mm dia. Rim)
1 x 50 mL (34 mm dia. RB)
10 x 1.6 to 7 mL
14 x 5 to 7 mL
2 x 50 mL
3 x 25 mL
3 x 50 mL (29 mm dia. FA)
3 x 50 mL (29 mm dia. RB)
5 x 25 mL
7 x 15 mL (17 mm dia. FA)
7 x 15 mL (17 mm dia. Rim)
8 x 10 mL
9 x 15 mL
Unit Size:
Pack of 2
Product Code. 15816752
Brand: Ohaus™ 30314915
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Type RB
Capacity 8 x 10 mL
Max. Tube Diameter 16 mm
For Use With (Equipment) R-S4x250/4MI Swing Out Rotors
Description Rack, 8x10ml D16mm RB, 2/pk
No. of Places 8
No. per Pack 2