Learn More
BOLA 2-Way Stopcocks
Product Code. 10558272
Change view
Click to view available options
Fits Tubing ID:
10 thread
4 thread
6 thread
8 thread
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fits Tubing ID Unit Size
10558272 6 thread Each
10359281 4 thread Each
10084960 8 thread Each
10185641 10 thread Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10558272 Supplier BOLA Supplier No. E65204
  • ONLINE EXCLUSIVE PRICE
    85.05€
    66.99€ / Pack of 1
    Save 18.06€
    21% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
2-Way Stopcocks PTFE, with glass-fibre reinforced PTFE nut
  • 2-way stopcock with straight bore and two connections or 3-way stopcock with T-shaped bore and three connections
  • With nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Conical stopcock plug, tightness is increased by turning the nut on the lower side.
  • 3-way stopcock plug with T-shaped mark of flow direction.
  • Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Autoclavable Autoclavable
Fits Tubing ID 6 thread
Closure Size M14
Certifications/Compliance FDA