Learn More
BOLA 2-Way Stopcocks
Click to view available options
Fits Tubing ID:
10 thread
4 thread
6 thread
8 thread
Unit Size:
Each
Description
- 2-way stopcock with straight bore and two connections or 3-way stopcock with T-shaped bore and three connections
- With nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
- Conical stopcock plug, tightness is increased by turning the nut on the lower side.
- 3-way stopcock plug with T-shaped mark of flow direction.
- Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Fits Tubing ID | 6 thread |
| Closure Size | M14 |
| Certifications/Compliance | FDA |