Learn More
BOLA 2-Way Stopcocks
Product Code. 10359281
Click to view available options
Fits Tubing ID:
10 thread
4 thread
6 thread
8 thread
Unit Size:
Each
Product Code. 10359281
Brand: BOLA E65202
2-Way Stopcocks PTFE, with glass-fibre reinforced PTFE nut
  • 2-way stopcock with straight bore and two connections or 3-way stopcock with T-shaped bore and three connections
  • With nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Conical stopcock plug, tightness is increased by turning the nut on the lower side.
  • 3-way stopcock plug with T-shaped mark of flow direction.
  • Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Autoclavable Autoclavable
Fits Tubing ID 4 thread
Closure Size M14
Certifications/Compliance FDA