Learn More
2-(Trifluoromethyl)phenylacetic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 3038-48-0 |
| Molecular Formula | C9H7F3O2 |
| MDL Number | MFCD00004327 |
| Quantity | 10 g |
| SMILES | O=C(O)CC1=CC=CC=C1C(F)(F)F |
| Molecular Weight (g/mol) | 204.15 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(Trifluoromethyl)phenylacetic acid |