Learn More
2-(Trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific™
Product Code. 11426870
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426870 5 g 5g
11416870 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426870 Supplier Thermo Scientific Alfa Aesar Supplier No. L19717.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1869-24-5
Molecular Formula C7H6F3NO2S
Molecular Weight (g/mol) 225.185
MDL Number MFCD00042420
InChI Key AFFPZJFLSDVZBV-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenesulfonamide, 2-trifluoromethyl benzene-1-sulfonamide, 2-trifluoromethyl benzenesulphonamide, o-trifluoromethylbenzenesulfonamide, benzenesulfonamide, 2-trifluoromethyl, pubchem11781, acmc-20a48s, ksc174g3h, 2-trifluoromethylbenzenesul fonamide
PubChem CID 2778018
IUPAC Name 2-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)N
Specifications
Melting Point 188°C to 192°C
Quantity 5 g
Beilstein 2697078
Formula Weight 225.19
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonamide