Learn More
2-(Trifluoromethyl)-3-ethoxydodecafluorohexane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Trifluoromethyl)-3-ethoxydodecafluorohexane |
| CAS | 297730-93-9 |
| Molecular Formula | C9H5F15O |
| Quantity | 10 g |
| Synonym | 3-Ethoxyperfluoro(2-methylhexane), HFE 7500, Novec 7500, Novec Engineered Fluid HFE 7500, Novec HFE 7500 |
| InChI Formula | InChI=1S/C9H5F15O/c1-2-25-6(15,3(10,7(16,17)18)8(19,20)21)4(11,12)5(13,14)9(22,23)24/h2H2,1H3 |
| SMILES | CCOC(F)(C(F)(F)C(F)(F)C(F)(F)F)C(F)(C(F)(F)F)C(F)(F)F |
| IUPAC Name | 4-ethoxy-1,1,1,2,2,3,3,4,5,6,6,6-dodecafluoro-5-(trifluoromethyl)hexane |
| Molecular Weight (g/mol) | 414.11 |
| Formula Weight | 414.0101 |