missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarbonyl Chloride, TRC

Product Code. 30454099
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454099 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454099 Supplier Toronto Research Chemicals Supplier No. TRCT36809050G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Thiophenecarbonyl Chloride
CAS 5271-67-0
Molecular Formula C5H3ClOS
Quantity 50 g
Synonym 2-(Chlorocarbonyl)thiophene, 2-(Chloroformyl)thiophene, 2-Thenoyl Chloride, 2-Thienylcarbonyl chloride, 2-Thiophencarbonyl chloride, 2-Thiophenecarboxylic Acid Chloride, 2-Thiophenecarboxylic Chloride, 2-Thiopheneformyl Chloride, 5-Thiophene-2-carbonyl Chloride, Thenoyl Chloride, Thiophen-2-ylcarbonyl Chloride, alpha-Thenoyl Chloride
InChI Formula InChI=1S/C5H3ClOS/c6-5(7)4-2-1-3-8-4/h1-3H
SMILES ClC(=O)c1cccs1
IUPAC Name thiophene-2-carbonyl chloride
Molecular Weight (g/mol) 146.59
Formula Weight 145.9593
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.