missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butyl) 3a-Methyl tetrahydropyrrolo[3,4-c]pyrrole-2,3a(1H,3H)-dicarboxylate, TRC

Product Code. 30423808
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423808 100 mg -
30420988 500 mg -
30411066 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423808 Supplier Toronto Research Chemicals Supplier No. TRCT185076100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1823265-38-8
Molecular Formula C13H22N2O4
Molecular Weight (g/mol) 270.32
IUPAC Name 5-O-tert-butyl 3a-O-methyl 1,2,3,4,6,6a-hexahydropyrrolo[3,4-c]pyrrole-3a,5-dicarboxylate
SMILES COC(=O)C12CNCC1CN(C2)C(=O)OC(C)(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H22N2O4/c1-12(2,3)19-11(17)15-6-9-5-14-7-13(9,8-15)10(16)18-4/h9,14H,5-8H2,1-4H3
Formula Weight 270.158
Chemical Name or Material 2-(tert-Butyl) 3a-Methyl tetrahydropyrrolo[3,4-c]pyrrole-2,3a(1H,3H)-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.