Learn More
2-(tert-Butoxycarbonyl)-5,7-dichloro-1,2,3,4-tetrahydroisoquinoline-6-carboxylic Acid, TRC
Product Code. 30336428
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336428 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30336428 Supplier Toronto Research Chemicals Supplier No. TRCB6941341G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(tert-Butoxycarbonyl)-5,7-dichloro-1,2,3,4-tetrahydroisoquinoline-6-carboxylic Acid
CAS 851784-82-2
Molecular Formula C15H17Cl2NO4
Quantity 1 g
Synonym 2-Boc-5,7-dichloro-1,2,3,4-tetrahydroisoquinoline-6-carboxylic Acid
InChI Formula InChI=1S/C15H17Cl2NO4/c1-15(2,3)22-14(21)18-5-4-9-8(7-18)6-10(16)11(12(9)17)13(19)20/h6H,4-5,7H2,1-3H3,(H,19,20)
SMILES CC(C)(C)OC(=O)N1CCc2c(Cl)c(C(=O)O)c(Cl)cc2C1
IUPAC Name 5,7-dichloro-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-6-carboxylic acid
Molecular Weight (g/mol) 346.21