Learn More
Ismatec™ 2-Stop Tubing, Tygon MHLL
Product Code. 10145072
Click to view available options
Tubing Size:
I.D. 0.38 mm
Unit Size:
Pack of 6
Product Code. 10145072
Brand: Ismatec™ SC0716
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Chemically resistant, long-life tubing with a smooth inner bore.
  • Chemically resistant to acetone, MEK, and other aggressive solvents.
  • Long-life tubing.
  • Plasticizer-free.
  • Smooth inner bore.
  • Low sorption maintains fluid integrity.
  • Minimal adhesion and diffusion.
Specifications
Certifications/Compliance FDA 21 CFR 177.2600;US Pharmacopoea Class VI
Color Code Orange-Green
Diameter (Metric) Inner 0.38 mm
Material Tygon MHLL
Overall Length (Metric) 381 mm
Min. Operating Temperature (Metric) −70°C
Packaging Quantity 6/Pk
Resistance Properties Acids: Excellent, Alkaline solutions: Excellent, Solvents: Excellent, Pressure: Not rec., Vacuum: Good, Viscous media: Good, Sterile media: Good
Tubing Size I.D. 0.38 mm
Max. Operating Temperature (Metric) 74°C