missing translation for 'onlineSavingsMsg'
Learn More

2-(quinolin-4-yl)ethan-1-amine, TRC

Product Code. 30370758
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370758 10 mg -
30373706 50 mg 50mg
30376151 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30370758 Supplier Toronto Research Chemicals Supplier No. TRCE59239510MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 860720-12-3
Molecular Formula C11 H12 N2
Molecular Weight (g/mol) 172.23
IUPAC Name 2-(4-quinolyl)ethanamine
SMILES NCCc1ccnc2ccccc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12N2/c12-7-5-9-6-8-13-11-4-2-1-3-10(9)11/h1-4,6,8H,5,7,12H2
Formula Weight 172.1
Chemical Name or Material 2-(Quinolin-4-yl)ethan-1-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.