Learn More
2-Pyrazine-d3-carboxylic Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Pyrazinecarboxylic Acid-d3 |
| CAS | 1794791-32-4 |
| Molecular Formula | C5 D3 H N2 O2 |
| Quantity | 0.1 g |
| Synonym | 3,5,6-Trideuteriopyrazine-2-carboxylic acid, Pyrazine-2-carboxylic Acid-3,5,6-D3, 2-Pyrazine-d3-carboxylic Acid, Pyrazinecarboxylic Acid-d3 |
| InChI Formula | InChI=1 S/C5H4N2O2/c8-5(9)4-3-6-1-2-7-4/h1-3 H,(H,8,9)/i1D,2 D,3 D |
| SMILES | [2 H]c1nc([2 H])c(nc1[2 H])C(=O)O |
| IUPAC Name | 3,5,6-trideuteriopyrazine-2-carboxylic acid |
| Molecular Weight (g/mol) | 127.116 |
| Formula Weight | 127.0461 g/mol |
| Show More |