missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpiperazine Dihydrochloride, TRC

Product Code. 30417392
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417392 500 mg -
30413256 50 mg -
30428346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417392 Supplier Toronto Research Chemicals Supplier No. TRCP399333500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872139-22-5
Molecular Formula C10H14N2 . 2(HCl)
Molecular Weight (g/mol) 235.15
IUPAC Name 2-phenylpiperazine;dihydrochloride
SMILES Cl.Cl.C1CNC(CN1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H14N2.2ClH/c1-2-4-9(5-3-1)10-8-11-6-7-12-10;;/h1-5,10-12H,6-8H2;2*1H
Formula Weight 234.0691
Chemical Name or Material 2-Phenylpiperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.