Learn More
2-Phenylglutaric Anhydride(>80%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Phenylglutaric Anhydride |
| CAS | 2959-96-8 |
| Molecular Formula | C11H10O3 |
| Quantity | 250 mg |
| Synonym | Dihydro-3-phenyl-2H-pyran-2,6(3H)-dione, 2-Phenylpentanedioic Acid Anhydride, 3-Phenyloxane-2,6-dione, NSC 65629 |
| InChI Formula | InChI=1S/C11H10O3/c12-10-7-6-9(11(13)14-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2 |
| SMILES | O=C1CCC(C(=O)O1)c2ccccc2 |
| IUPAC Name | 3-phenyloxane-2,6-dione |
| Molecular Weight (g/mol) | 190.19 |
| Formula Weight | 190.063 |