Learn More
2-Phenylbutyronitrile, 95%
Product Code. 11488966
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488966 50 g 50g
11478966 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11488966 Supplier Thermo Scientific Alfa Aesar Supplier No. A17093.18
Request Bulk or Custom Format
CAS: 769-68-6 | C10H11N | 145.205 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 769-68-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00001876
InChI Key IZPUPXNVRNBDSW-UHFFFAOYSA-N
Synonym 2-phenylbutyronitrile, 2-phenylbutanentrile, 1-cyano-1-phenylpropane, butanenitrile, 2-phenyl, benzeneacetonitrile, .alpha.-ethyl, butyronitrile, 2-phenyl, benzeneacetonitrile, ?-ethyl, .alpha.-phenylbutyronitrile, alpha-phenylbutyronitrile, dl-2-phenylbutyronitrile
PubChem CID 95334
IUPAC Name 2-phenylbutanenitrile
SMILES CCC(C#N)C1=CC=CC=C1
Specifications
Density 0.975
Boiling Point 102°C to 104°C (7 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.5086
Quantity 50 g
UN Number UN3276
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 2-Phenylbutyronitrile