missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-(pyridin-2-yl)ethan-1-amine, TRC

Product Code. 30424145
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424145 100 mg 100mg
30415286 1 g -
30420607 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424145 Supplier Toronto Research Chemicals Supplier No. TRCP205451100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95898-97-8
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.26
IUPAC Name 2-phenyl-2-pyridin-2-ylethanamine
SMILES NCC(c1ccccc1)c2ccccn2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H14N2/c14-10-12(11-6-2-1-3-7-11)13-8-4-5-9-15-13/h1-9,12H,10,14H2
Formula Weight 198.1157
Chemical Name or Material 2-Phenyl-2-(pyridin-2-yl)ethan-1-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.