missing translation for 'onlineSavingsMsg'
Learn More

2-Oxo-Zoniporide Hydrochloride, TRC

Product Code. 30417728
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417728 Supplier Toronto Research Chemicals Supplier No. TRCO87046510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Oxo-Zoniporide Hydrochloride
CAS 372078-42-7
Purity Grade Notes HPLC
Molecular Formula C17 H16 N6 O2 . HCl
Quantity 10 mg
Synonym N-(Aminoiminomethyl)-5-cyclopropyl-1-(1,2-dihydro-2-oxo-5-quinolinyl)-1H-pyrazole-4-carboxamide Hydrochloride
InChI Formula InChI=1S/C17H16N6O2.ClH/c18-17(19)22-16(25)11-8-20-23(15(11)9-4-5-9)13-3-1-2-12-10(13)6-7-14(24)21-12;/h1-3,6-9H,4-5H2,(H,21,24)(H4,18,19,22,25);1H
SMILES O=C(N1)C=CC(C1=CC=C2)=C2N3C(C4CC4)=C(C(NC(N)=N)=O)C=N3.Cl
IUPAC Name N-carbamimidoyl-5-cyclopropyl-1-(2-oxo-1,2-dihydroquinolin-5-yl)-1H-pyrazole-4-carboxamide hydrochloride
Molecular Weight (g/mol) 372.81
Formula Weight 372.11
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.