Learn More
2-O-(Alpha-D-Galactopyranosyl)-D-galactose, TRC
Product Code. 30179380
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179380 10 mg 10mg
30179566 1 mg 1mg
30178937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179380 Supplier Toronto Research Chemicals Supplier No. TRCG15470010MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93601-68-4
Molecular Formula C12 H22 O11
Molecular Weight (g/mol) 342.3
IUPAC Name (3R,4S,5R,6R)-6-(hydroxymethyl)-3-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-2,4,5-triol
SMILES OC[C@H]1OC(O)[C@H](O[C@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H]1O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C12H22O11/c13-1-3-6(16)8(18)10(11(20)21-3)23-12-9(19)7(17)5(15)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5+,6+,7+,8+,9-,10-,11?,12-/m1/s1
Formula Weight 342.1162
Chemical Name or Material 2-O-(a-D-Galactopyranosyl)-D-galactose