missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrotoluene-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Nitrotoluene-d7 |
| CAS | 84344-04-7 |
| Molecular Formula | C7 2H7 N O2 |
| Quantity | 1 g |
| Synonym | 2-Nitrotoluene-d7 |
| InChI Formula | InChI=1 S/C7H7NO2/c1-6-4-2-3-5-7(6)8(9)10/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(c1[2 H])[N+](=O)[O-])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-5-nitro-6-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 144.18 |
| Formula Weight | 144.0916 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction