missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl bromide, 98+%

Product Code. 11454484
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11454484

Brand: Thermo Scientific Alfa Aesar A13127.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3958-60-9 | C7H6BrNO2 | 216.034 g/mol

2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with alcohols, bases, amines and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3958-60-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007184
InChI Key HXBMIQJOSHZCFX-UHFFFAOYSA-N
Synonym 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
PubChem CID 77569
IUPAC Name 1-(bromomethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CBr)[N+](=O)[O-]

Specifications

Melting Point 45°C to 48°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 638991
Solubility Information Insoluble in water.
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.