missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-5-phenylthioaniline, TRC

Product Code. 30466604
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466604 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466604 Supplier Toronto Research Chemicals Supplier No. TRCN49699010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitro-5-phenylthioaniline
CAS 43156-47-4
Molecular Formula C12H10N2O2S
Quantity 10 g
Synonym 2-Nitro-5-(phenylthio)-benzenamine, 1-Amino-2-nitro-5-phenylthiobenzene, 2-Amino-4-phenylthio-1-nitrobenzene
InChI Formula InChI=1S/C12H10N2O2S/c13-11-8-10(6-7-12(11)14(15)16)17-9-4-2-1-3-5-9/h1-8H,13H2
SMILES Nc1cc(Sc2ccccc2)ccc1[N+](=O)[O-]
IUPAC Name 2-nitro-5-phenylsulfanylaniline
Molecular Weight (g/mol) 246.28
Formula Weight 246.0463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.