missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-thiocyanato Aniline, TRC

Product Code. 30468782
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468782 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468782 Supplier Toronto Research Chemicals Supplier No. TRCN5653201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Nitro-4-thiocyanato Aniline
CAS 54029-45-7
Molecular Formula C7H5N3O2S
Quantity 1 g
Synonym 4-Amino-3-nitrophenyl Thiocyanate, 2-Nitro-4-thiocyanatobenzenamine, 4-Thiocyanato-2-nitroaniline.
InChI Formula InChI=1S/C7H5N3O2S/c8-4-13-5-1-2-6(9)7(3-5)10(11)12/h1-3H,9H2
SMILES Nc1ccc(SC#N)cc1[N+](=O)[O-]
IUPAC Name (4-amino-3-nitrophenyl) thiocyanate
Molecular Weight (g/mol) 195.2
Formula Weight 195.0102
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.