missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1-heptanol, 98%

Product Code. p-7033264
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11439520

Brand: Thermo Scientific Alfa Aesar H50340.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10042-59-8 | C10H22O | 158.285 g/mol

2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

Solubility
Insoluble in water. Miscible with most common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10042-59-8
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00046768
InChI Key YLQLIQIAXYRMDL-UHFFFAOYSA-N
Synonym 2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302
PubChem CID 24847
IUPAC Name 2-propylheptan-1-ol
SMILES CCCCCC(CCC)CO

Specifications

Quantity 1 g
Solubility Information Insoluble in water. Miscible with most common organic solvents.
Formula Weight 158.28
Percent Purity 98%
Chemical Name or Material 2-n-Propyl-1-heptanol
compliance-icons
Product Identifier
  • 2-n-Propyl-1-heptanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-n-Propyl-1-heptanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.