missing translation for 'onlineSavingsMsg'
Learn More

2-n-Butoxyethyl acetate, 98%

Product Code. 11357466
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 11357466

Brand: Thermo Scientific Alfa Aesar L09515.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-07-2 | C8H16O3 | 160.21 g/mol

2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Butoxyethyl acetate is used as a solvent for nitrocellulose lacquers, enamels, coating additives, epoxy resins, and varnishes. It is a film coalescing aid for polyvinyl acetate latex. Further, it stimulates the release of prostaglandin E(2), an arachidonic acid metabolite, in human epidermal keratinocytes. In addition to this, it serves as an ingredient in ink removers and spot removers.

Solubility
Miscible with hydrocarbons and organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-07-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00009457
InChI Key NQBXSWAWVZHKBZ-UHFFFAOYSA-N
Synonym butoxyethyl acetate, butyl glycol acetate, butylglycol acetate, ethylene glycol monobutyl ether acetate, 2-butoxyethanol acetate, butyl cellosolve acetate, ektasolve eb acetate, ethanol, 2-butoxy-, acetate, butylcelosolvacetat, 1-acetoxy-2-butoxyethane
PubChem CID 8160
IUPAC Name 2-butoxyethyl acetate
SMILES CCCCOCCOC(C)=O

Specifications

Melting Point -64°C
Density 0.942
Boiling Point 191°C to 192°C
Flash Point 71°C (159°F)
Odor Fruit-like
Linear Formula CH3COOCH2CH2O(CH2)3CH3
Refractive Index 1.413
Quantity 500 mL
Beilstein 1756960
Solubility Information Miscible with hydrocarbons and organic solvents. Slightly miscible with water.
Formula Weight 160.21
Percent Purity 98%
Chemical Name or Material 2-n-Butoxyethyl acetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.