Learn More
2-Morpholinobenzaldehyde, 97%, Thermo Scientific™
Product Code. 10019471
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
10 g
missing translation for 'unitSize'
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10019471 1 g 1g
10321925 5 g 5g
10374912 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Product Code. 10019471 missing translation for 'mfr' Thermo Scientific Maybridge missing translation for 'supplierNo' SEW05383DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 58028-76-5
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00662562
InChI Key GTTAEWVBVHSDLX-UHFFFAOYSA-N
Synonym 2-morpholinobenzaldehyde, 2-morpholin-4-yl-benzaldehyde, 2-4-morpholino benzaldehyde, 2-morpholin-4-yl benzaldehyde, benzaldehyde, 2-4-morpholinyl, morpholinobenzaldehyde, pubchem16276, acmc-1b05v, 2-morpholinobenzenecarbaldehyde, 4-2-formylphenyl morpholine
PubChem CID 2737010
IUPAC Name 2-morpholin-4-ylbenzaldehyde
SMILES O=CC1=CC=CC=C1N1CCOCC1
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 2-Morpholinobenzaldehyde