missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole-d5 (Major), TRC

Product Code. 30483032
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483032 100 mg 100mg
30482289 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30483032 Supplier Toronto Research Chemicals Supplier No. TRCM312987100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697806-98-7
Molecular Formula C4HD5N2
Molecular Weight (g/mol) 87.13
Synonym 1H-2-Methylimidazole-d5, 2-Methyl-1H-imidazole-d5, 2MZ-d5, 2MZ-H-d5, 2MZ-PW-d5, Actiron 2MI-d5, Curezol 2MZ-d5, Curezol 2MZ-P-d5, Denka CN 25-d5, Epicure MI 2-d5, Epikure MI 2-d5, Imicure AMI 2-d5, NSC 21394-d5
SMILES [2H]C1=C(N=C(N1)C([2H])([2H])[2H])[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H6N2/c1-4-5-2-3-6-4/h2-3H,1H3,(H,5,6)/i1D3,2D,3D
Chemical Name or Material 2-Methylimidazole-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.