Learn More
(2-Methylenecyclopropyl)methanol, TRC
Specifications
Specifications
| Chemical Name or Material | (2-Methylenecyclopropyl)methanol |
| CAS | 29279-66-1 |
| Molecular Formula | C5H8O |
| Quantity | 100 mg |
| Synonym | 2-Methylenecyclopropanemethanol |
| InChI Formula | InChI=1S/C5H8O/c1-4-2-5(4)3-6/h5-6H,1-3H2 |
| SMILES | OCC1CC1=C |
| IUPAC Name | (2-methylidenecyclopropyl)methanol |
| Molecular Weight (g/mol) | 84.12 |
| Formula Weight | 84.0575 |