missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzimidazole, 98%

Product Code. 11413527
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11413527

Brand: Thermo Scientific Alfa Aesar A17290.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-15-6 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-15-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005598
InChI Key LDZYRENCLPUXAX-UHFFFAOYSA-N
PubChem CID 11984
SMILES CC1=NC2=CC=CC=C2N1

Specifications

Melting Point 174°C to 178°C
Quantity 250 g
Beilstein 112264
Synonym 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole
IUPAC Name 2-methyl-1H-benzimidazole
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 2-Methylbenzimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.