Learn More
2-Methyl-N-methyltetrahydro-2-furanmethanamine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-N-methyltetrahydro-2-furanmethanamine |
| CAS | 7179-95-5 |
| Molecular Formula | C7H15NO |
| Quantity | 25 mg |
| Synonym | Tetrahydro-N,2-dimethyl-2-furanmethanamine |
| InChI Formula | InChI=1S/C7H15NO/c1-7(6-8-2)4-3-5-9-7/h8H,3-6H2,1-2H3 |
| SMILES | CC1(CNC)CCCO1 |
| IUPAC Name | N-methyl-1-(2-methyltetrahydrofuran-2-yl)methanamine |
| Molecular Weight (g/mol) | 129.2 |
| Formula Weight | 129.12 |