Learn More
2-Methyl-d3-propyl Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-1-propanol-3,3,3-D3 |
| CAS | 95927-04-1 |
| Molecular Formula | C4 D3 H7 O |
| Quantity | 0.5 g |
| Synonym | 1-Propan-3,3,3-d3-ol, 2-methyl- (9 CI), 2-Methyl-d3-propyl alcohol, 3,3,3-Trideuterio-2-methylpropan-1-ol, 2-Methyl-1-propan-3,3,3-d3-ol, Isobutanol-d3, Isobutyl-d3 alcohol, 2-(Methyl-d3)-1-propanol, 2-Methyl-1-propanol-3,3,3-D3 |
| InChI Formula | InChI=1 S/C4H10O/c1-4(2)3-5/h4-5 H,3H2,1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C)CO |
| IUPAC Name | 3,3,3-trideuterio-2-methylpropan-1-ol |
| Molecular Weight (g/mol) | 77.1401 |
| Formula Weight | 77.092 g/mol |
| Show More |