missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-d3-propyl-3,3,3-d3 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-d3-propyl-3,3,3-d3 Alcohol |
| CAS | 72182-69-5 |
| Molecular Formula | C4 D6 H4 O |
| Quantity | 0.5 g |
| Synonym | 1-Propan-3,3,3-d3-ol, 2-(methyl-d3)- (9 CI), 3,3,3-Trideuterio-2-(trideuteriomethyl)propan-1-ol, 2-(Methyl-d3)-1-propan-3,3,3-d3-ol, 2-Methyl-d3-propyl-3,3,3-d3 alcohol, Isobutanol-d6, Isobutyl-d6 alcohol, 2-Methyl-1-propanol-d6, 2-Methyl-1-propanol-D6 (dimethyl-D6) |
| InChI Formula | InChI=1 S/C4H10O/c1-4(2)3-5/h4-5 H,3H2,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(CO)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,3-trideuterio-2-(trideuteriomethyl)propan-1-ol |
| Molecular Weight (g/mol) | 80.1586 |
| Formula Weight | 80.1108 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction