Learn More
2-Methyl-d3-propionic-3,3,3-d3 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Isobutyric acid dimethyl-d6 |
| CAS | 29054-08-8 |
| Molecular Formula | C4 2H6 H2 O2 |
| Quantity | 0.5 g |
| Synonym | Propanoic-3,3,3-d3 acid, 2-(methyl-d3)-, Isobutyric-3,3,3,3',3',3'-d6 acid (8 CI), 2-(Methyl-d3)propanoic-3,3,3-d3 acid, Isobutyric acid D6 (dimethyl-D6), Isobutyric acid-d6 |
| InChI Formula | InChI=1 S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid |
| Molecular Weight (g/mol) | 94.14 |
| Formula Weight | 94.0901 g/mol |
| Show More |