missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-d3-3-butyn-1,1,1-d3-2-ol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-3-butyn-2-ol-D6 (dimethyl-D6) |
| CAS | 57444-27-6 |
| Molecular Formula | C5 D6 H2 O |
| Quantity | 0.25 g |
| Synonym | 3-Butyn-1,1,1-d3-2-ol, 2-(methyl-d3)- (9 CI, ACI), 2-(Methyl-d3)-3-butyn-1,1,1-d3-2-ol (ACI), 1,1,1-Trideutero-2-(trideuteromethyl)-3-butyn-2-ol, 1,1,1-Trideutero-2-trideutermethyl-3-butyn-2-ol, 2-Methyl-3-butyn-2-ol-d6, 2-Methyl-3-butyn-2-ol-D6 (dimethyl-D6), 2-Methyl-d3-3-butyn-1,1,1-d3-2-ol |
| InChI Formula | InChI=1 S/C5H8O/c1-4-5(2,3)6/h1,6 H,2-3H3/i2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)(C#C)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1-trideuterio-2-(trideuteriomethyl)but-3-yn-2-ol |
| Molecular Weight (g/mol) | 90.1534 |
| Formula Weight | 90.0952 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction