Learn More
2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic Acid, TRC
Product Code. 30470444
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470444 100 mg 100mg
30480785 10 mg 10mg
30478046 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30470444 Supplier Toronto Research Chemicals Supplier No. TRCM219195100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 117724-63-7
Molecular Formula C6H4NO2F3S
Molecular Weight (g/mol) 211.16
Synonym 2-Methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic Acid, 2-Methyl-4-trifluoromethylthiazole-5-carboxylic Acid
IUPAC Name 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid
SMILES Cc1nc(c(s1)C(=O)O)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H4F3NO2S/c1-2-10-4(6(7,8)9)3(13-2)5(11)12/h1H3,(H,11,12)
Formula Weight 210.9915
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic Acid