Learn More
2-Methyl-3-phenylpyrazine, TRC
Product Code. 30136573
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136573 500 mg 500mg
30132943 1 g 1g
30009328 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136573 Supplier Toronto Research Chemicals Supplier No. TRCM356433500MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29444-53-9
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.21
IUPAC Name 2-methyl-3-phenylpyrazine
SMILES Cc1nccnc1c2ccccc2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C11H10N2/c1-9-11(13-8-7-12-9)10-5-3-2-4-6-10/h2-8H,1H3
Formula Weight 170.0844
Chemical Name or Material 2-Methyl-3-phenylpyrazine