Learn More
2-Methyl-2-propoxypropane, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-propoxypropane |
| CAS | 29072-93-3 |
| Molecular Formula | C7H16O |
| Quantity | 100 mg |
| Synonym | 2-Methyl-2-propoxypropane, 1,1-Dimethylethyl Propyl Ether, 1-tert-Butoxypropane, Propyl 1,1-Dimethylethyl Ether, Propyl tert-Butyl Ether, n-Propyl tert-Butyl Ether, tert-Butyl n-Propyl Ether, tert-Butyl Propyl Ether |
| InChI Formula | InChI=1S/C7H16O/c1-5-6-8-7(2,3)4/h5-6H2,1-4H3 |
| SMILES | CCCOC(C)(C)C |
| IUPAC Name | 2-methyl-2-propoxypropane |
| Molecular Weight (g/mol) | 116.2 |
| Formula Weight | 116.1201 |