missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-(4-(methylthio)phenyl)-2-morpholinopropan-1-one, TRC

Product Code. 30479241
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479241 2.5 g 2.5g
30464411 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479241 Supplier Toronto Research Chemicals Supplier No. TRCM3396252.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-methyl-4'-(methylthio)-2-Morpholinopropiophenone
CAS 71868-10-5
Purity Grade Notes HPLC
Molecular Formula C15H21NO2S
Quantity 2.5 g
Synonym 2-Methyl-1-[4-(methylthio)phenyl]-2-(4-morpholinyl)-1-propanone, 2-Methyl-1-(4-methylsulfanylphenyl)-2-morpholinopropan-1-one, 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholino-1-propanone, 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholinopropane-1-one, 2-Methyl-1-[4-(methylthio)phenyl]-2-morpholinopropanone, 2-Methyl-4'-(methylthio)-2-morpholinopropiophenone, 2-Methyl[4-(methylthio)phenyl]-2-morpholinopropan-1-one, Caccure 907, Chemcure 709, Ciba 907, D 177, GR 907, Genocure PMP, I 907, IC 907, IHT-PI 907, IR 907, Irg 907, Irgacure 184C, Irgacure 907, Irgacure I 907, JR 907, Photocure 907, Photoinitiator 907, Runtecure 1107, SB-PI 777, UV-Cure D 177, photoinitiator 970, α-Methyl-4-(methylmercapto)-mopholinopropiophenone, α-Methyl-4-(methylmercapto)-α-morpholinopropiophenone
InChI Formula InChI=1S/C15H21NO2S/c1-15(2,16-8-10-18-11-9-16)14(17)12-4-6-13(19-3)7-5-12/h4-7H,8-11H2,1-3H3
SMILES CSc1ccc(cc1)C(=O)C(C)(C)N2CCOCC2
IUPAC Name 2-methyl-1-(4-methylsulfanylphenyl)-2-morpholin-4-ylpropan-1-one
Molecular Weight (g/mol) 279.4
Formula Weight 279.1293
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.