Learn More
2-Methyl-1,4-butanediol, TRC
Product Code. 30475290
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475290 10 mg 10mg
30475047 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30475290 Supplier Toronto Research Chemicals Supplier No. TRCM22249510MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-1,4-butanediol
CAS 2938-98-9
Molecular Formula C5H12O2
Quantity 10 mg
InChI Formula InChI=1S/C5H12O2/c1-5(4-7)2-3-6/h5-7H,2-4H2,1H3
SMILES C(CCO)(CO)C
IUPAC Name 2-methylbutane-1,4-diol
Molecular Weight (g/mol) 104.15
Formula Weight 104.0837