missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-bis(trimethylsiloxy)benzene, 98%, Thermo Scientific™

Product Code. 11499627
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499627 5 g 5g
11409637 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499627 Supplier Thermo Scientific Alfa Aesar Supplier No. B22116.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78018-53-8
Molecular Formula C13H24O2Si2
Molecular Weight (g/mol) 268.50
MDL Number MFCD02683083
InChI Key IPYOHMJSEPFQSQ-UHFFFAOYSA-N
Synonym 2-methyl-1,4-bis trimethylsiloxy benzene, trimethyl 3-methyl-4-trimethylsilyl oxy phenoxy silane, 2,5-bis trimethylsiloxy toluene, methylhydroquinone, 2tms derivative, methylhydroquinone, bis trimethylsilyl ether, 1,4-bis trimethylsilyl oxy-2-methylbenzene, benzene,2-methyl-1,4-bis trimethylsilyl oxy, 2-methyl-1,4-phenylene bis oxy bis trimethylsilane
PubChem CID 12691803
IUPAC Name trimethyl-(2-methyl-4-trimethylsilyloxyphenoxy)silane
SMILES CC1=C(O[Si](C)(C)C)C=CC(O[Si](C)(C)C)=C1

Specifications

Quantity 5 g
Formula Weight 268.51
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material 2-Methyl-1,4-bis(trimethylsiloxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.