missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11424460
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424460 1 g 1g
11414460 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424460 Supplier Thermo Scientific Alfa Aesar Supplier No. H50060.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532391-31-4
Molecular Formula C12H18BNO3
Molecular Weight (g/mol) 235.09
MDL Number MFCD05663507
InChI Key JEJYNUCGRAHMDO-UHFFFAOYSA-N
Synonym 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-methoxypyridine-3-boronic acid pinacol ester, 2-methoxypyridin-3-ylboronic acid pinacol ester, 2-methoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pubchem16545, ksc498e8t, 2-methoxypyridine-3-boronic acid, pinacol ester, 2-methoxypyridin-3-yl boronic acid pinacol ester
PubChem CID 17750166
IUPAC Name 2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES COC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 235.09
Percent Purity 97%
Chemical Name or Material 2-Methoxypyridine-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.