Learn More
2-Methoxy-5-methylphenyl isothiocyanate, 99%, Thermo Scientific™
Product Code. 11347536
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347536 1 g 1g
11357536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11347536 Supplier Thermo Scientific Alfa Aesar Supplier No. L11085.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 190774-56-2
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041083
InChI Key CYKFAZDMLSXWKZ-UHFFFAOYSA-N
Synonym 2-methoxy-5-methylphenyl isothiocyanate, 2-methoxy-5-methylphenylisothiocyanate, benzene,2-isothiocyanato-1-methoxy-4-methyl, acmc-20anrv, 2-methoxy-5-methylbenzenisothiocyanate, 2-isothiocyanato-1-methoxy-4-methyl-benzene, #, benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
PubChem CID 601672
IUPAC Name 2-isothiocyanato-1-methoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)N=C=S
Specifications
Density 1.146
Boiling Point 288°C
Flash Point >110°C (230°F)
Refractive Index 1.63
Quantity 1 g
UN Number UN2922
Beilstein 6919550
Sensitivity Moisture sensitive
Formula Weight 179.24
Percent Purity 99%
Chemical Name or Material 2-Methoxy-5-methylphenyl isothiocyanate