Learn More
2-Mercaptoethanol-d6, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Mercaptoethanol-D6 |
| CAS | 203645-37-8 |
| Molecular Formula | C2 D6 O S |
| Quantity | 0.1 g |
| Synonym | Ethan-1,1,2,2-d4-ol-d, 2-(mercapto-d)- (9 CI), 2-Mercaptoethanol-D6, 2-Mercaptoethanol-d6, 2-(Mercapto-d)ethan-1,1,2,2-d4-ol-d, 2-Mercaptoethanol-O,S,1,1,2,2-d6, 2-Sulfanylethanol-d6, β-Hydroxyethanethiol-d6 |
| InChI Formula | InChI=1 S/C2H6OS/c3-1-2-4/h3-4 H,1-2H2/i1D2,2D2,3 D/hD |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])S[2 H] |
| IUPAC Name | 1,1,2,2-tetradeuterio-1-deuteriooxy-2-deuteriosulfanylethane |
| Molecular Weight (g/mol) | 84.1704 |
| Formula Weight | 84.0516 g/mol |
| Show More |