Learn More
2-Isopropylphenyl isocyanate, 97%
Product Code. p-7045414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392896 1 g 1g
11302906 5 g 5g
11312906 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11392896 Supplier Thermo Scientific Alfa Aesar Supplier No. L10618.03
Request Bulk or Custom Format
CAS: 56309-56-9 | C10H11NO | 161.204 g/mol
2-Isopropylphenyl isocyanate is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropylphenyl isocyanate is used as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56309-56-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00013850
InChI Key MWBSQVPLYJZPTG-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
PubChem CID 91797
IUPAC Name 1-isocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=O
Specifications
Boiling Point 66°C to 67°C (5 mmHg)
Flash Point 87°C (188°F)
Quantity 1 g
UN Number UN2206
Beilstein 3588928
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylphenyl isocyanate