missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodopropane-1,1,1,3,3,3-d6, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Iodopropane-1,1,1,3,3,3-D6 |
| CAS | 39091-64-0 |
| Molecular Formula | C3 D6 H I |
| Quantity | 1 g |
| Synonym | Propane-1,1,1,3,3,3-d6, 2-iodo- (6 CI, 9 CI), 1,1,1,3,3,3-Hexadeutero-2-iodopropane, 2-Iodopropane-1,1,1,3,3,3-d6, 2-Iodo-1,1,1,3,3,3-hexadeuteriopropane, 2-Iodo-1,1,1,3,3,3-hexadeuteropropane, 2-Iodopropane-1,1,1,3,3,3-D6, 2-Iodopropane-d6 |
| InChI Formula | InChI=1 S/C3H7I/c1-3(2)4/h3H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(I)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,3,3,3-hexadeuterio-2-iodopropane |
| Molecular Weight (g/mol) | 176.0291 |
| Formula Weight | 175.9969 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction